Learn More
Spectrum™ Chemical Albumin, Bovine Serum, Fraction V, Heat-Shock Isolation, BiotechGrade
SDP
Catalog No. 18600353 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-353 1 kg Poly Bottle
18-600-351 25 g Poly Bottle
18-600-352 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-600-353 Supplier Spectrum™ Chemical Supplier No. A36111KGBL
Add to Cart
Add to Cart
A3611, 9048-46-8
Spectrum™ Chemical Albumin, Bovine Serum, Fraction V, Heat-Shock Isolation, BiotechGrade is a serum albumin protein that comes from cows. It is also known as Bovine serum albumin and usually used as a protein concentration standard in laboratory experiments. Spectrum offers highly pure reagents suitable for biochemical research and analysis.
Specifications
Quantity 1 kg
Packaging Poly Bottle
Form Lyophilized