Learn More
Alpco Ltd Canine Albumin ELISA
SDP
Supplier:  Alpco Ltd 41ALBCAE01
Encompass
The Canine Albumin ELISA is a highly sensitive two-site enzyme linked immunoassay for measuring albumin in serum, tissue extracts and other biological fluids of dogs.
Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3569