Learn More
Sigma Aldrich Fine Chemicals Biosciences Albumin chicken egg w250MG
Catalog No. 502898747
Encompass
Ovalbumin an egg white protein is produced in the oviduct of hen. It has 3 sites of post synthetic modification and an N-terminal acetyl group. This product is free of S-ovalbumin a heat stable form of egg albumin formed when eggs are stored.
Catalog No. 50-289-8747 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A7641250MG
Ovalbumin an egg white protein is produced in the oviduct of hen. It has 3 sites of post synthetic modification and an N-terminal acetyl group. This product is free of S-ovalbumin a heat stable form of egg albumin formed when eggs are stored.