Learn More
Invent Biotechnologies Inc ALBUMIN DEPLETION REAGENT 20ML
Catalog No. NC2389197
Encompass
Human/animal serum and plasma are the best sources of samples for biomarker identification. However, the presence of albumin in large amounts interferes with many assays and functional proteomic analysis. Serum albumin accounts for 50-70% of the total serum protein that causes loss of resolution for 1D and 2D electrophoresis. Effective removal of albumin using our reagent can increase sensitivity of the assay for many folds. Unlike a resin-based depletion kit (that only deals with microliter amounts of a sample), Minute albumin depletion reagent can handle a few microliters and up to several milliliters of samples. Over 90% of albumin can be removed in less than 5 minutes.
Catalog No. NC2389197 Supplier Invent Biotechnologies Inc Supplier No. WA013
Human/animal serum and plasma are the best sources of samples for biomarker identification. However, the presence of albumin in large amounts interferes with many assays and functional proteomic analysis. Serum albumin accounts for 50-70% of the total serum protein that causes loss of resolution for 1D and 2D electrophoresis. Effective removal of albumin using our reagent can increase sensitivity of the assay for many folds. Unlike a resin-based depletion kit (that only deals with microliter amounts of a sample), Minute albumin depletion reagent can handle a few microliters and up to several milliliters of samples. Over 90% of albumin can be removed in less than 5 minutes.