Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albumin, Egg, Powder, Purified, J.T. Baker™

Catalog No. 02002211 Shop All Avantor Products
Change view
Click to view available options
Quantity:
125 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-211 125 g Poly Bottle
02-002-212 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-002-211 Supplier Avantor Supplier No. 044004
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 9006-50-2
Molecular Formula C6H8N2O4
Molecular Weight (g/mol) 172.14
InChI Key QCVGEOXPDFCNHA-UHFFFAOYSA-N
Synonym tridodeoylamine
PubChem CID 25022239
IUPAC Name 5,5-dimethyl-2,4-dioxo-1,3-oxazolidine-3-carboxamide
SMILES CC1(C(=O)N(C(=O)O1)C(=O)N)C

Specifications

Packaging Poly Bottle
Quantity 125 g
Grade Purified
Physical Form Powder
Chemical Name or Material Albumin, Egg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.