Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Albumin, from Eggs
SCI_ED

Catalog No. S25132B Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Reagent Grade
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25132B 500 g Reagent Grade
S25132 25 g Reagent Grade
S25132A 100 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S25132B
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 9006-59-1
Molecular Formula C16H26O5
Molecular Weight (g/mol) 298.38
MDL Number MFCD00130424
InChI Key UOXVFQCRPDLSFN-UHFFFAOYNA-N
Synonym 3,4-dihydroxy-2-methoxy-4-methyl-3-2-methyl-3-3-methyl-but-2-enyl-oxiranyl-cyclohexanone, ova, 2s,3r,4r-3,4-dihydroxy-2-methoxy-4-methyl-3-2s,3r-2-methyl-3-3-methylbut-2-en-1-yl oxiran-2-yl cyclohexan-1-one, 2s,3r,4r-3,4-dihydroxy-2-methoxy-4-methyl-3-2s,3r-2-methyl-3-3-methylbut-2-enyl oxiran-2-yl cyclohexan-1-one
PubChem CID 5289086
IUPAC Name 3,4-dihydroxy-2-methoxy-4-methyl-3-[2-methyl-3-(3-methylbut-2-en-1-yl)oxiran-2-yl]cyclohexan-1-one
SMILES COC1C(=O)CCC(C)(O)C1(O)C1(C)OC1CC=C(C)C

Specifications

Color Yellow
Density 1g/mL
Boiling Point 140°F
Packaging Reagent Grade
Quantity 500 g
Formula Weight 33.8 to 46.1
Percent Purity 98%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Albumin, from Eggs
Safety handling Check Alert storage code Red
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.