Learn More
Sigma Aldrich Fine Chemicals Biosciences Albumin from chicken egg w10G
Catalog No. 502903879
Encompass
Chicken egg albumin is the major protein constituent of egg whites. Chicken egg albumin is a phosphorylated-glycoprotein. from the amino acid sequence the peptide portion of the molecule consists of 385 residues and has a molecular weight of 42.7 kDa.
Catalog No. 50-290-3879 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A537810G
Chicken egg albumin is the major protein constituent of egg whites. Chicken egg albumin is a phosphorylated-glycoprotein. from the amino acid sequence the peptide portion of the molecule consists of 385 residues and has a molecular weight of 42.7 kDa.