Learn More
Sigma Aldrich Fine Chemicals Biosciences Albumin from chicken egg w250G
Catalog No. 502899415
Encompass
Ovalbumin make 75% of the total egg white protein.
Catalog No. 50-289-9415 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A5253250G
Ovalbumin make 75% of the total egg white protein.