Learn More
Sigma Aldrich Fine Chemicals Biosciences Albumin from chicken egg w25G
Catalog No. 502903833
Encompass
Chicken egg albumin is the major protein constituent of egg whites. Chicken egg albumin is a phosphorylated-glycoprotein. from the amino acid sequence the peptide portion of the molecule consists of 385 residues and has a molecular weight of 42.7 kDa.
Catalog No. 50-290-3833 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A537825G
Chicken egg albumin is the major protein constituent of egg whites. Chicken egg albumin is a phosphorylated-glycoprotein. from the amino acid sequence the peptide portion of the molecule consists of 385 residues and has a molecular weight of 42.7 kDa.