Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Albumin, Human, 30% Solution

Catalog No. 0210371250 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-103-71250 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-103-71250 Supplier MP Biomedicals™ Supplier No. 0210371250
Only null left
Add to Cart
Add to Cart

Albumin, Human, 30% Solution is tested negative for the presence of Hepatitis B Surface Antigen (HBsAg), HIV I/II, HIV-1Ag, HCV, ALT and Syphilis.

  • Regulates the colloidal osmotic pressure of blood.
  • Solubilizes lipids.
  • Used as a blocking agent.
  • CAS Number: 70024-90-7
TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 188.02 g/mol
Quantity 50 mL
Source Human
Biological Activity Human serum albumin undergoes three different post-translational modifications: oxidation, glycation, and S-nitrosylation. Modifications usually occur on the surface of the globular protein, and do not significantly affect conformation. However, modification strongly affects binding of fatty acids and drug molecules.
Form Liquid
Melting Point -147.6°C
Boiling Point -36.6°C
Density 1.352 at 20° (liquid)
Vapor Pressure 6.63X10+3 mm Hg at 25°C
Solubility Soluble in water, 5.7 mg/L at 15°C
Synonym Human Serum Albumin, Albumin from human serum, HAS
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.