Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Albumin, Human, 96%

Catalog No. AAJ6678003
Click to view available options
Quantity:
500 mg
1 g
5 g

Albumin, CAS # 70024-90-7, is derived from human serum and is a cell membrane protein involved in facilitated diffusion and active transport of substances, including steroids, fatty acids, and thyroid hormones.

Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones, and is vital in regulating the colloidal osmotic pressures of blood. Albumin is also seen to bind to exogenous substances, particularly drugs (e.g., ibuprofen, warfarin), and strongly influence their pharmacokinetics. Oxidative stress leading to changes in the redox state of albumin has widely varied effects on its physiological function. Albumin-bound fluorescence was used in serum of patients with chronic renal failure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Albumin is derived from human serum

• Albumin is affected by oxidative stress that can alter its redox state and thereby alter its physiological function

Application

• Albumin has been used in the preparation of nanocarriers as advanced drug delivery systems

• It has the ability to facilitate diffusion and active transport of substances such as steroids, fatty acids, and thyroid hormones

• It also helps maintain the colloidal osmotic pressures of blood and buffers pH

• It can bind to some substances, including drugs such as ibuprofen and warfarin, and strongly influence their pharmacokinetics

• It is involved in the formulation of media for culture of mammalian cells for laboratory research

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Protein Carrier, Cell Transport
Formulation Lyophilized powder
Quantity 1 g
Source Human Serum
Storage Requirements 2° to 8°C
Regulatory Status RUO
Product Type Albumin, Human, 96%
Recombinant Native
Form Lyophilized
Purity or Quality Grade 96%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.