Learn More
Cayman Chemical Albumin (human) ELISA Monoclonal Antibody, 100 dtn
SDP
Catalog No. 502942299
Encompass_Preferred
Albumin (human) ELISA Monoclonal Antibody has been tested and formulated to work exclusively with Cayman’s Albumin (human) ELISA Kit (Item No. 501760).
Catalog No. 50-294-2299 Supplier Cayman Chemical Supplier No. 401762100 DTN
Albumin (human) ELISA Monoclonal Antibody has been tested and formulated to work exclusively with Cayman’s Albumin (human) ELISA Kit (Item No. 501760).