Learn More
Athens Research And Technology Inc ALBUMIN HUMAN PL 1GM
Supplier:  Athens Research And Technology Inc 16160112021GR
Encompass_Preferred
Albumin, Human Plasma: 1 GRAM; Salt-free lyophilized solid; MW 66,000; Extinction Coefficient = 0.58; Purity: > =95% by SDS-PAGE. This native protein purified in the Athens Research & Technology laboratory. Molecular Biology Reagent, Enzyme
Catalog No. 50-489-595