Learn More
Sigma Aldrich Fine Chemicals Biosciences Albumn-fluorescen soth250MG
Catalog No. 502905018
Encompass
Albumin makes up 60% of the plasma protein which is produced by the liver. It regulates colloid osmotic pressure. Albumin is implicated in the transportation distribution and metabolism of fatty acids thyroid hormones and drugs. It reduces weight gain blocks pulmonary edema decreases ascites and lowers tissue edema.
Catalog No. 50-290-5018 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A9771250MG
Albumin makes up 60% of the plasma protein which is produced by the liver. It regulates colloid osmotic pressure. Albumin is implicated in the transportation distribution and metabolism of fatty acids thyroid hormones and drugs. It reduces weight gain blocks pulmonary edema decreases ascites and lowers tissue edema.