Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Albuterol Sulfate, USP, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18600358 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-358 5 g Amber Glass Bottle
18-600-359 25 g Amber Glass Bottle
18-600-360 100 g Amber Glass Bottle
18-600-361 1 kg Amber Glass Bottle
18-600-362 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-600-358 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AL1565GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

AL156, 51022-70-9, (C13H21NO3)2·H2SO4

Spectrum™ Chemical Albuterol Sulfate, USP is a bronchodilator used to treat or prevent bronchospasm in people with reversible obstructive airway disease. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 51022-70-9
Molecular Formula C26H44N2O10S
Molecular Weight (g/mol) 576.70
InChI Key BNPSSFBOAGDEEL-UHFFFAOYNA-N
IUPAC Name bis(4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol); sulfuric acid
SMILES OS(O)(=O)=O.CC(C)(C)NCC(O)C1=CC=C(O)C(CO)=C1.CC(C)(C)NCC(O)C1=CC=C(O)C(CO)=C1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 98.5 to 101%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.