Learn More
Kyfora Bio ALC-0159 25 MG
Supplier:  Kyfora Bio 1449225
Encompass_Preferred
ALC-0159 (N,N-dimyristylamide of 2-hydroxyacetic acid) is a PEGylated lipid commonly utilized in the formulation of lipid nanoparticles (LNPs) for the delivery of mRNA and other therapeutic agents. PEGylated lipids like ALC-0159 enhance the stability, circulation time, and bioavailability of LNPs in vivo, making them essential for effective delivery in mRNA vaccine development and other nucleic acid-based applications. This reagent is intended exclusively for research purposes.Applications: Lipid nanoparticles (LNPs), mRNA vaccine development
Catalog No. 50-255-9809