Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Avanti Polar Lipids ALC-0159, Methoxypolyethylene glycol conjugated lipid, 25 mg powder, Cationic Lipid, PEG-2000

Supplier:  Avanti Polar Lipids 880155P25MG

Encompass

ALC-0159 is a PEGylated lipid designed to enhance lipid nanoparticle (LNP) drug delivery systems, significantly improving the bioavailability of therapeutics. By forming a hydrophilic shield, it reduces immune recognition and extends systemic circulation. This excipient is crucial for mRNA-based vaccines, enabling efficient encapsulation and intracellular delivery, with applications in gene therapy and advanced pharmaceutical uses such as CRISPR and siRNA delivery.

  • Cationic lipid and polymerizable lipid
  • Empirical formula: C121H243NO47
  • Molecular weight: 2464.20 Da
  • Purity: >99%
  • Form: Powder
  • PEG moiety: PEG-2000
  • Application: mRNA-based vaccines, nucleic acid delivery, gene therapy
  • Storage temperature: -20°C
  • Shipping: Dry ice

Catalog No. NC2373065


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.