Learn More
Medchemexpress LLC HY-138170 100mg , ALC-0315 CAS:2036272-55-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY138170100MG
Encompass_Preferred
HY-138170 100mg ALC-0315 CAS: 2036272-55-4 ALC-0315 is an ionisable aminolipid that is responsible for mRNA compaction and aids mRNA cellular delivery and its cytoplasmic release through suspected endosomal destabilization. ALC-0315 can be used to form lipid nanoparticle (LNP) delivery vehicles. Lipid-Nanoparticles have been used in the research of mRNA COVID-19 vaccine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9046