Learn More
Cayman Chemical ALC-0315 nalogous-1 10mg
SDP
Catalog No. 502918971
Encompass_Preferred
An ALC-0315 derivative; has been used in the synthesis of ionizable cationic lipids used in the generation of LNPs
Catalog No. 50-291-8971 Supplier Cayman Chemical Supplier No. 3859110 MG
An ALC-0315 derivative; has been used in the synthesis of ionizable cationic lipids used in the generation of LNPs