Learn More
Echelon Biosciences Research Labs ALC-0315 TRAILBLAZER KT PREMIX
SDP
Catalog No. NC3343891
Encompass
LNP Trailblazer Kits are designed for quick assembly and assessment of RNA-LNPs in biological systems. Each kit includes a buffer for preparation of RNA cargo and the necessary lipids to form lipid nanoparticles. Once formed RNA-LNPs are stable in neutral pH buffers for several weeks allowing them to be used serially for repeat experiments. The kit components are compatible with bench top microfluidic mixers and hand mixing by pipette.
Catalog No. NC3343891 Supplier Echelon Biosciences Research Labs Supplier No. K6300MALC0315
LNP Trailblazer Kits are designed for quick assembly and assessment of RNA-LNPs in biological systems. Each kit includes a buffer for preparation of RNA cargo and the necessary lipids to form lipid nanoparticles. Once formed RNA-LNPs are stable in neutral pH buffers for several weeks allowing them to be used serially for repeat experiments. The kit components are compatible with bench top microfluidic mixers and hand mixing by pipette.