Learn More
Cayman Chemical Alcaftadn 10mg
SDP
Catalog No. 502985521
Encompass_Preferred
A H1 histamine receptor antagonist (IC50 = 8.6 nM) with anti-inflammatory and mast cell stabilizing effects; reduces edema and erythema in a guinea pig model of allergic conjunctivitis (ED50 = 0.1 mg/kg); has been used in clinical formulations in eye drops for treatment of ocular itching associated with allergic conjunctivitis
Catalog No. 50-298-5521 Supplier Cayman Chemical Supplier No. 2129010 MG
A H1 histamine receptor antagonist (IC50 = 8.6 nM) with anti-inflammatory and mast cell stabilizing effects; reduces edema and erythema in a guinea pig model of allergic conjunctivitis (ED50 = 0.1 mg/kg); has been used in clinical formulations in eye drops for treatment of ocular itching associated with allergic conjunctivitis