Learn More
NOVOZYMES BIOLOGICAL INC 211 Alcalase 2.4L FG, Food grade
Catalog No. NC3705119
Encompass
This item has a minimum qty of 25 per supplier requirements.
Sustine 211 is a serine endopeptidase that performs stereoselective hydrolysis of amino esters, selective esters, and amides.Proteases are enzymes that catalyze the proteolysis into smaller polypeptides or amino acid units at increased reaction rates. They cleave peptide bonds through hydrolysis, catalyzing stereoselective hydrolysis of amino esters and selective esters to synthesize new molecules in an environmentally friendly way. Unparalleled regio- and enantioselectivity Faster reaction rates under mild reaction conditions Optimal temperature range: 30°-65° C Optimal pH range: 7-9 Works well with di- and poly-esters Food grade
Catalog No. NC3705119 Supplier NOVOZYMES BIOLOGICAL INC Supplier No. SUSTINE 211
This item has a minimum qty of 25 per supplier requirements.
Sustine 211 is a serine endopeptidase that performs stereoselective hydrolysis of amino esters, selective esters, and amides.Proteases are enzymes that catalyze the proteolysis into smaller polypeptides or amino acid units at increased reaction rates. They cleave peptide bonds through hydrolysis, catalyzing stereoselective hydrolysis of amino esters and selective esters to synthesize new molecules in an environmentally friendly way. Unparalleled regio- and enantioselectivity Faster reaction rates under mild reaction conditions Optimal temperature range: 30°-65° C Optimal pH range: 7-9 Works well with di- and poly-esters Food grade