Learn More
ACROBiosystems ACROBiosystems Human ALCAM Protein, Fc Tag
Supplier:  ACROBiosystems CD6H52591MG
Encompass_Preferred
Recombinant Protein;1MG;Human ALCAM, Fc Tag (CD6-H5259) is expressed from human 293 cells (HEK293). It contains AA Trp 28 - Ala 526 (Accession # NP_001618).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 82.6 kDa. The protein migrates as 95-116 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7518