Learn More
ACROBiosystems Recombinant Protein;Human ALCAM Protein, Fc Tag;HEK293;200UG;CD6,CD166,ALCAM,CD6L,KG-CAM,MEMD
Catalog No. 501659066
Encompass_Preferred
Recombinant Protein;Human ALCAM Protein, Fc Tag;200UG;Human ALCAM, Fc Tag (CD6-H5259) is expressed from human 293 cells (HEK293). It contains AA Trp 28 - Ala 526 (Accession # NP_001618).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 82.6 kDa. The protein migrates as 95-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-9066 Supplier ACROBiosystems Supplier No. CD6H5259200UG
Recombinant Protein;Human ALCAM Protein, Fc Tag;200UG;Human ALCAM, Fc Tag (CD6-H5259) is expressed from human 293 cells (HEK293). It contains AA Trp 28 - Ala 526 (Accession # NP_001618).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 82.6 kDa. The protein migrates as 95-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.