Learn More
ACROBiosystems Recombinant Protein;Human ALCAM Protein;HEK293;1MG;CD6,CD166,ALCAM,CD6L,KG-CAM,MEMD
Catalog No. 501658147
Encompass_Preferred
Recombinant Protein;Human ALCAM Protein;1MG;Human ALCAM, His Tag (CD6-H5228) is expressed from human 293 cells (HEK293). It contains AA Trp 28 - Ala 526 (Accession # Q13740-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 57.8 kDa. As a result of glycosylation, the protein migrates as 70-100 kDa under reducing (R) condition, and 66-100 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-165-8147 Supplier ACROBiosystems Supplier No. CD6H52281MG
Recombinant Protein;Human ALCAM Protein;1MG;Human ALCAM, His Tag (CD6-H5228) is expressed from human 293 cells (HEK293). It contains AA Trp 28 - Ala 526 (Accession # Q13740-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 57.8 kDa. As a result of glycosylation, the protein migrates as 70-100 kDa under reducing (R) condition, and 66-100 kDa under non-reducing (NR) condition (SDS-PAGE).