Learn More
Volu Sol Alcoholic Formalin 10% Neutral Buffered, 5 gal / 20 L, Volu-Sol™
SDP
Catalog No. 501694710 Shop All Volu Sol Products
Encompass_Preferred
Volu-Sol?s Alcoholic Formalin 10% Neutral Buffered is an excellent fixative for use on frozen sections. This custom blend performs well with fatty or thick tissue sections and can penetrate up to three (3) times faster than typical 10% Neutral Buffered Formalin solutions.
This item is not returnable. View return policy
Catalog No. 50-169-4710 Supplier Volu Sol Supplier No. VFA640
Volu-Sol?s Alcoholic Formalin 10% Neutral Buffered is an excellent fixative for use on frozen sections. This custom blend performs well with fatty or thick tissue sections and can penetrate up to three (3) times faster than typical 10% Neutral Buffered Formalin solutions.