Learn More
GA International Alcohol And Water Resistant Cryogenic Marker - Fine Tip Green 6/pack
Catalog No. NC1293554
Encompass_Preferred
Alcohol and water-resistant cryogenic marker for use in various laboratory applications. Available in black or green, these fine tip permanent markers are provided in packs of 6 markers. Compatible with all major cryogenic labels from GA International, the marker provides an alcohol-resistant marking on most surfaces.
Catalog No. NC1293554 Supplier GA International Supplier No. SM1G6
Alcohol and water-resistant cryogenic marker for use in various laboratory applications. Available in black or green, these fine tip permanent markers are provided in packs of 6 markers. Compatible with all major cryogenic labels from GA International, the marker provides an alcohol-resistant marking on most surfaces.