Learn More
Newcomer Supply Alcian Blue Stain 1%, pH 2.5 Aqueous, 500 ml
SDP
Catalog No. NC1736651
Encompass
Newcomer Supply Alcian Blue Stain 1%, pH 2.5 Aqueous is designed to stain acid epithelial mucins (sialomucin, sulfomucin) as well as stromal (mesenchymal) mucin in the Alcian Blue Stain 1%, pH 2.5 procedure. Use: Acidic Mucins & Carboxylated Mucosubstances; including Barrett’s Esophageal Biopsy. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
Catalog No. NC1736651 Supplier Newcomer Supply Supplier No. 1003B
Newcomer Supply Alcian Blue Stain 1%, pH 2.5 Aqueous is designed to stain acid epithelial mucins (sialomucin, sulfomucin) as well as stromal (mesenchymal) mucin in the Alcian Blue Stain 1%, pH 2.5 procedure. Use: Acidic Mucins & Carboxylated Mucosubstances; including Barrett’s Esophageal Biopsy. Shelf Life is 1 year from date of manufacture. Store at 15-30°C.