Learn More
Newcomer Supply Alcian Blue 1%, pH 2.5 Stain Kit, 250 ml kit
SDP
Catalog No. NC2048335
Encompass
Newcomer Supply Alcian Blue 1%, pH 2.5 Stain Kit procedure, with included hyaluronidase method for stromal mucin digestion, is designed to stain acid epithelial mucin (sialomucin, sulfomucin) as well as stromal (mesenchymal) mucin. The hyaluronidase digestion step is used for further differentiation of acid epithelial from stromal mucin.
Catalog No. NC2048335 Supplier Newcomer Supply Supplier No. 9102A
Newcomer Supply Alcian Blue 1%, pH 2.5 Stain Kit procedure, with included hyaluronidase method for stromal mucin digestion, is designed to stain acid epithelial mucin (sialomucin, sulfomucin) as well as stromal (mesenchymal) mucin. The hyaluronidase digestion step is used for further differentiation of acid epithelial from stromal mucin.