Learn More
Electron Microscopy Sciences Alcian Blue 1% 250 ML
SDP
Catalog No. 5031752
Encompass_Preferred
Used in Alcian Blue Method (pH 2.5) for staining weakly acidic sulfated mucosubstances, hyaluronic acid and, sialomucins. See AFIP Manual of Histological Stains Methods, 3rd Ed, Ed. L. Luna: New York McGraw Hill Publications, c. 1968, p. 163. or Lev, R. & Spicer. S.S., J. Histochem Cystochem, 12:309, 1964
Catalog No. 50-317-52 Supplier Electron Microscopy Sciences Supplier No. 2632301
Used in Alcian Blue Method (pH 2.5) for staining weakly acidic sulfated mucosubstances, hyaluronic acid and, sialomucins. See AFIP Manual of Histological Stains Methods, 3rd Ed, Ed. L. Luna: New York McGraw Hill Publications, c. 1968, p. 163. or Lev, R. & Spicer. S.S., J. Histochem Cystochem, 12:309, 1964