Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcian blue 8 GX (C.I.74240), For Microscopy, Certistain™, MilliporeSigma™

Catalog No. M1052340010 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1052340010 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1052340010 Supplier MilliporeSigma Supplier No. 1.05234.0010

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 33864-99-2
Molecular Formula C56H68Cl4CuN16S4
Molecular Weight (g/mol) 1298.86
MDL Number MFCD00010720
InChI Key XRWQTDIELZDCPS-UHFFFAOYSA-J
PubChem CID 17749099
IUPAC Name (9Z,18Z,26Z,35Z)-4,14,22,32-tetrakis({[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}methyl)-9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11(16),12,14,17(41),18,20,22,24,26,28(40),29(34),30,32,35-nonadecaene-38,38-bis(ylium) tetrachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].CN(C)C(SCC1=CC2=C(C=C1)\C1=N\C3=C4C=CC(CSC(N(C)C)=[N+](C)C)=CC4=C4\N=C5/N=C(/N=C6\N([Cu++]N34)\C(=N/C2=N1)C1=CC(CSC(N(C)C)=[N+](C)C)=CC=C61)C1=C5C=C(CSC(N(C)C)=[N+](C)C)C=C1)=[N+](C)C

Specifications

Odor Odorless
Packaging Glass bottle
Color Dark Blue
Quantity 10 g
Physical Form Powder
Formula Weight 1298.88
Chemical Name or Material Alcian blue 8 GX
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.