Learn More
Medchemexpress LLC Alcian Blue 8GX | 33864-99-2 | 77% | 1 ML
SDP
Catalog No. 5000410134
Encompass_Preferred
Alcian Blue 8GX is a widely used phthalocyanine dye that binds to glycoproteins and glycosaminoglycans. It has a broad range of applications in biological staining, including the staining of proteins in brain tumors and DNA in cells and tissues.
  • Binds to glycoproteins and glycosaminoglycans.
  • Used for biological staining of proteins in brain tumors and DNA in cells and tissues.
  • Applicable in Alcian Blue 8GX working solution preparation.
  • Utilized for Alcian Blue 8GX staining of cell micromass cultures.
  • Effective for Alcian Blue 8GX staining of glycosaminoglycans.
  • Used in Alcian Blue 8GX staining of fetal cartilage.
  • Can be used for protein determination based on its resonance light scattering enhancement effect.
Catalog No. 50-004-10134 Supplier Medchemexpress LLC Supplier No. HYD000110MM/1ML
Alcian Blue 8GX is a widely used phthalocyanine dye that binds to glycoproteins and glycosaminoglycans. It has a broad range of applications in biological staining, including the staining of proteins in brain tumors and DNA in cells and tissues.
  • Binds to glycoproteins and glycosaminoglycans.
  • Used for biological staining of proteins in brain tumors and DNA in cells and tissues.
  • Applicable in Alcian Blue 8GX working solution preparation.
  • Utilized for Alcian Blue 8GX staining of cell micromass cultures.
  • Effective for Alcian Blue 8GX staining of glycosaminoglycans.
  • Used in Alcian Blue 8GX staining of fetal cartilage.
  • Can be used for protein determination based on its resonance light scattering enhancement effect.