Learn More
Abcam Alcian Blue 8G x , Cationic dye, 5G
Catalog No. 502705360
Encompass_Preferred
MW 1298.9 Da, Purity >50%. Cationic dye. Forms insoluble complexes with glycosaminoglycans in solution. Stains interphase nuclei, especially mitotic figures.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5360 Supplier Abcam Supplier No. AB1452505G
MW 1298.9 Da, Purity >50%. Cationic dye. Forms insoluble complexes with glycosaminoglycans in solution. Stains interphase nuclei, especially mitotic figures.
The product is subject to the following: Abcam Restricted Use Statement