Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcian Blue 8GX, Certified Grade, Sigma-Aldrich

Catalog No. 01259049 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-049 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-259-049 Supplier MilliporeSigma Supelco Supplier No. A315710G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alcian Blue 8GX has been used for the staining of sulfated proteoglycans in murine MPC (mesenchymal progenitor cell) nodule. It has been used for the staining of sulfated proteoglycans in bovine articular chondrocytes.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Alcian Blue 8GX
CAS 33864-99-2
Color Dark Blue
Quantity 10 g
Molecular Weight (g/mol) 1298.86 g/mol
Grade Certified Biological Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.