Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcian Blue 8GX, Laboratory Grade Sigma-Aldrich

Catalog No. 01258941 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-941 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-941 Supplier MilliporeSigma Supelco Supplier No. 0550010G
Add to Cart
Add to Cart
  • Alcian Blue 8GX (Cl 74240), also known as Alcian Blue 8GN or Ingrain Blue 1, is a cationic, copper phthalocyanine dye
  • Four cationic guanidinium groups are attached to the ring system via thioether links, which form an insoluble pigment when broken by hydroxide ions or nucleophiles
  • The dye is basic, tetravalent, and blue in color due to its copper content
  • It forms salt linkages with the acid groups of acid mucopolysaccharides.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Alcian Blue
CAS 33864-99-2
Solubility Information Methanol: Water (1:1): 0.01 g/10 mL
Molecular Weight (g/mol) 1298.86 g/mol
Color Blue/Green
Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.