Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcian Blue 8GX, MP Biomedicals™

Catalog No. MP215262425 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215262425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215262425 Supplier MP Biomedicals, Inc Supplier No. 0215262425
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Shows ring structure with amidinothiomethyl groups. Alcian Blue 8GX, MP Biomedicals is a cationic dye used to quantitatively determine glycosaminoglycans by forming insoluble complexes in solution. The dye increases in selectivity when increasing amounts

Alcian Blue 8GX increases in selectivity when increasing amounts of magnesium chloride are added. In contrast, studies show that when electrolyte concentration is very low, it has trouble binding to carboxylate or phosphate groups.

  • Quantitatively determine glycosaminoglycans by forming insoluble complexes in solution
  • Determine proteins based on their Resonance Light Scattering by strongly enhancing the signals
  • Useful as a bacterial stain and as a dye for histiocytes and fibroblasts
  • Used as a heteroglycan stain for neutral, sulfated and phosphated mucopolysaccharides and glycosaminoglycans in tissues such as cartilage and extracellular matrices
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75881-23-1
Molecular Formula C56H68Cl4CuN16S4
Molecular Weight (g/mol) 1298.858
InChI Key CKLBXIYTBHXJEH-UHFFFAOYSA-J
Synonym Alcian Blue, Ingrain Blue 1
PubChem CID 16211091
SMILES CN(C)C(=[N+](C)C)SCC1=CC2=C(C=C1)C3=NC4=NC(=NC5=C6C=C(C=CC6=C([N-]5)N=C7C8=C(C=CC(=C8)CSC(=[N+](C)C)N(C)C)C(=N7)N=C2[N-]3)CSC(=[N+](C)C)N(C)C)C9=C4C=C(C=C9)CSC(=[N+](C)C)N(C)C.[Cl-].[Cl-].[Cl-].[Cl-].[Cu+2]

Specifications

Solubility Information Soluble in absolute ethanol, cellosolve, ethylene glycol, water (8mg/mL) or ethanol (1mg/mL); practically insoluble in xylene
Quantity 25 g
Physical Form Powder
Formula Weight 1298.88
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.