Learn More
Newcomer Supply Alcian Blue 8GX, Powder, 5 g
SDP
Catalog No. NC9895486
Encompass
CI 74240. Shelf Life is 4 years from date of manufacture. Store at 15-30°C.
Catalog No. NC9895486 Supplier Newcomer Supply Supplier No. 1002A
CI 74240. Shelf Life is 4 years from date of manufacture. Store at 15-30°C.