Learn More
Description
Alcian Blue 8GX, also known as Ingrain Blue 1, is a basic copper phthalocyanine dye whose copper content imparts a blue color. It is tetravalent, cationic, and water-soluble in nature. It forms salt linkages with the acid groups of acid mucopolysaccharides.
Specifications
Specifications
| Chemical Name or Material | Alcian Blue |
| CAS | 33864-99-2 |
| Color | Blue |
| Quantity | 10 g |
| Molecular Weight (g/mol) | 1298.86 g/mol |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.