Learn More
Epredia™ Alcian Blue/PAS Special Stain Kit
GSA_VA
Catalog No. 23900664 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Alcian Blue/PAS Special Stain Kit
Catalog No. Product Type
23-900-664 Alcian Blue/PAS Special Stain Kit
1 options
This item is not returnable. View return policy
Catalog No. 23-900-664 Supplier Epredia™ Supplier No. 87023
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Demonstrate the presence of both neutral and acidic mucosubstances with the Epredia™ Alcian Blue/PAS Special Stain Kit
  • Can be used alone to stain acid mucins and other carboxylated and sulfated acid mucosubstances blue on tissue sections
  • Periodic Acid Schiff (PAS) reaction is used to stain glycogen, basement membranes and neutral mucosubstances
Includes: Alcian Blue pH 2.5 stain solution, periodic acid solution (0.5%) and Schiff reagent
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Product Type Alcian Blue/PAS Special Stain Kit
WARNING: Cancer - www.P65Warnings.ca.gov