Learn More
Newcomer Supply Alcian Blue/PAS (Periodic Acid Schiff) Stain Kit, 500 ml kit
SDP
Catalog No. NC2102254
Encompass
Newcomer Supply Alcian Blue/PAS Stain Kit procedure is used to differentiate acidic epithelial mucins (sialomucin, sulfomucin), neutral epithelial mucins and stromal (mesenchymal) mucins and is a means of detecting the overall presence of mucins.
This item is not returnable. View return policy
Catalog No. NC2102254 Supplier Newcomer Supply Supplier No. 91022B
Newcomer Supply Alcian Blue/PAS Stain Kit procedure is used to differentiate acidic epithelial mucins (sialomucin, sulfomucin), neutral epithelial mucins and stromal (mesenchymal) mucins and is a means of detecting the overall presence of mucins.