Learn More
Diagnostic Biosystems ALCIAN BLUE PH 10 Alcian Blue, pH 1.0
Catalog No. 502407341
Encompass_Preferred
This kit stains strongly sulfated mucosubstances. Strongly sulfated mucosub-stances stain blue in paraffin tissue sections, while the nuclei stain pink to red and the cytoplasm and background stain pale pink.
Catalog No. 50-240-7341 Supplier Diagnostic Biosystems Supplier No. KT002
This kit stains strongly sulfated mucosubstances. Strongly sulfated mucosub-stances stain blue in paraffin tissue sections, while the nuclei stain pink to red and the cytoplasm and background stain pale pink.