Learn More
Newcomer Supply Alcian Blue pH 2.5, Goblet Cell Control Slides, 98/set
SDP
Catalog No. NC1742633
Encompass
Newcomer Supply Alcian Blue pH 2.5, Goblet Cell Control Slides are for the positive histochemical staining of goblet cells as well as other acid epithelial mucins (sialomucin, sulfomucin).  Goblet cells normally line the small intestine but are an abnormal finding in Barrett’s esophagus. Alcian Blue pH 2.5, Goblet Cell Control Slides contain sections of positive staining small intestine.
Catalog No. NC1742633 Supplier Newcomer Supply Supplier No. 4051B
Newcomer Supply Alcian Blue pH 2.5, Goblet Cell Control Slides are for the positive histochemical staining of goblet cells as well as other acid epithelial mucins (sialomucin, sulfomucin).  Goblet cells normally line the small intestine but are an abnormal finding in Barrett’s esophagus. Alcian Blue pH 2.5, Goblet Cell Control Slides contain sections of positive staining small intestine.