Learn More
Scytek Laboratories Alcian Blue (pH 2.5) Staining Kit: Large
SDP
Catalog No. NC1969824
Encompass_Preferred
The Alcian Blue (pH 2.5) Stain Kit is intended for use in the histological visualization of sulfated and carboxylated acid mucopolysaccharides and sulfated and carboxylated sialomucins (glycoproteins).Acidic Sulfated Mucosubstances: BlueHyaluronic Acid: BlueSialomucins: BlueNuclei: RedBackground: PinkControl: Intestine or Colon.Kit contents: AAG125, Acetic Acid Solution (3%), 125 ml x 2.ANC250, Alcian Blue Solution, pH 2.5, 250 ml.PAQ250, Periodic Acid Solution (1%), 250 ml.SRF250, Schiff’s Solution, 250 ml.HMM125, Hematoxylin, Mayer’s (Lillie’s), 125 ml x 2.
This item is not returnable. View return policy
Catalog No. NC1969824 Supplier Scytek Laboratories Supplier No. AFR1
The Alcian Blue (pH 2.5) Stain Kit is intended for use in the histological visualization of sulfated and carboxylated acid mucopolysaccharides and sulfated and carboxylated sialomucins (glycoproteins).Acidic Sulfated Mucosubstances: BlueHyaluronic Acid: BlueSialomucins: BlueNuclei: RedBackground: PinkControl: Intestine or Colon.Kit contents: AAG125, Acetic Acid Solution (3%), 125 ml x 2.ANC250, Alcian Blue Solution, pH 2.5, 250 ml.PAQ250, Periodic Acid Solution (1%), 250 ml.SRF250, Schiff’s Solution, 250 ml.HMM125, Hematoxylin, Mayer’s (Lillie’s), 125 ml x 2.