Learn More
Scytek Laboratories Alcian Blue (pH 2.5) Staining Kit: Small
SDP
Catalog No. NC1969825
Encompass_Preferred
The Alcian Blue (pH 2.5) Stain Kit is intended for use in the histological visualization of sulfated and carboxylated acid mucopolysaccharides and sulfated and carboxylated sialomucins (glycoproteins).Acidic Sulfated Mucosubstances: BlueHyaluronic Acid: BlueSialomucins: BlueNuclei: RedBackground: PinkControl: Intestine or Colon.Kit contents: AAG030, Acetic Acid Solution (3%), 30 ml.ANC030, Alcian Blue Solution, pH 2.5, 30 ml.PAQ030, Periodic Acid Solution (1%), 30 ml.SRF030, Schiff’s Solution, 30 ml. HMM030, Hematoxylin, Mayer’s (Lillie’s), 30 ml.
This item is not returnable. View return policy
Catalog No. NC1969825 Supplier Scytek Laboratories Supplier No. AFR2
The Alcian Blue (pH 2.5) Stain Kit is intended for use in the histological visualization of sulfated and carboxylated acid mucopolysaccharides and sulfated and carboxylated sialomucins (glycoproteins).Acidic Sulfated Mucosubstances: BlueHyaluronic Acid: BlueSialomucins: BlueNuclei: RedBackground: PinkControl: Intestine or Colon.Kit contents: AAG030, Acetic Acid Solution (3%), 30 ml.ANC030, Alcian Blue Solution, pH 2.5, 30 ml.PAQ030, Periodic Acid Solution (1%), 30 ml.SRF030, Schiff’s Solution, 30 ml. HMM030, Hematoxylin, Mayer’s (Lillie’s), 30 ml.