Learn More
Diagnostic Biosystems ALCIAN BLUE PH 25 Alcian Blue, pH 2.5
Catalog No. 502407411
Encompass_Preferred
The Alcian Blue (pH 2.5) Stain Kit is intended for use in the histological visualization of sulfated and carboxylated acid mucopolysaccharides and sulfated and carboxylated sialomucins (glycoproteins).
Catalog No. 50-240-7411 Supplier Diagnostic Biosystems Supplier No. KT003
The Alcian Blue (pH 2.5) Stain Kit is intended for use in the histological visualization of sulfated and carboxylated acid mucopolysaccharides and sulfated and carboxylated sialomucins (glycoproteins).