Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alcian Blue, Pyridine Variant, Sigma-Aldrich

Catalog No. 01258949 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-949 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-949 Supplier MilliporeSigma Supelco Supplier No. 33563025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used in hematology, histology, and diagnostic assay manufacturing.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Alcian Blue, pyridine variant
CAS 123439-83-8
Color Blue
Quantity 25 g
Molecular Weight (g/mol) 1086.36 g/mol
Percent Purity ≥85%
Grade Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.