Learn More
Ihc World LLC ALCIAN BLUE STAIN KIT
SDP
Catalog No. NC0432666
Encompass_Preferred
Kit is designed to stain formalin fixed, paraffin embedded tissue sections, & may be used for frozen sections as well. Strongly acidic mucosubstances will be stained blue, nuclei will be stained pink to red, & cytoplasm will be stained pale pink.
Catalog No. NC0432666 Supplier Ihc World LLC Supplier No. IW3000
Kit is designed to stain formalin fixed, paraffin embedded tissue sections, & may be used for frozen sections as well. Strongly acidic mucosubstances will be stained blue, nuclei will be stained pink to red, & cytoplasm will be stained pale pink.