Learn More
Medchemexpress LLC Alclofenac | 22131-79-9 | 99.4% | 226.66 | 250 MG
SDP
Catalog No. 5000366412
Encompass_Preferred
Alclofenac (Allopydin) is a non-steroidal anti-inflammatory agent (NSAID). It acts as a prostaglandin H2 synthase inhibitor. It can be used for research related to rheumatoid arthritis, degenerative joint disease, and ankylosing spondylitis.
  • Acts as a non-steroidal anti-inflammatory agent (NSAID)
  • Functions as a prostaglandin H2 synthase inhibitor
  • Suitable for research on rheumatoid arthritis, degenerative joint disease, and ankylosing spondylitis
Catalog No. 50-003-66412 Supplier Medchemexpress LLC Supplier No. 250MGHYW009706
Alclofenac (Allopydin) is a non-steroidal anti-inflammatory agent (NSAID). It acts as a prostaglandin H2 synthase inhibitor. It can be used for research related to rheumatoid arthritis, degenerative joint disease, and ankylosing spondylitis.
  • Acts as a non-steroidal anti-inflammatory agent (NSAID)
  • Functions as a prostaglandin H2 synthase inhibitor
  • Suitable for research on rheumatoid arthritis, degenerative joint disease, and ankylosing spondylitis