Learn More
Cayman Chemical Alclometasone Dipropionate, Purity: ≥98%, CAS: 66734-13-2, 10 mg
SDP
Catalog No. 502994371
Encompass_Preferred
A corticosteroid; reduces cutaneous anaphylaxis reactions induced by tuberculin or albumin in mice when administered topically (20 μl of a 0.1% solution); inhibits androgen-dependent cytochrome P450 activity and the O-depropylation activity of 7-alkoxy-coumarin O-dealkylase in a dose-dependent manner in male rats; has no effect on hepatic drug metabolism in female rats or mice of both sexes
Catalog No. 50-299-4371 Supplier Cayman Chemical Supplier No. 2390110 MG
A corticosteroid; reduces cutaneous anaphylaxis reactions induced by tuberculin or albumin in mice when administered topically (20 μl of a 0.1% solution); inhibits androgen-dependent cytochrome P450 activity and the O-depropylation activity of 7-alkoxy-coumarin O-dealkylase in a dose-dependent manner in male rats; has no effect on hepatic drug metabolism in female rats or mice of both sexes