Learn More
MSC Zep Instant Hand Sanitizer Gel, Alcohol-Based, 1200 mL
Catalog No. 502837599
Encompass
Zep Instant Hand Sanitizer Gel is an alcohol-based hand sanitizer designed for effective hygiene without the need for water or towels. This 1200 mL gel helps to control many microorganisms of public health concern and can be used repeatedly throughout the day without irritation. It quickly and completely evaporates from the skin, leaving no residue, eliminating the need for towels.
  • May be used repeatedly without irritation
  • Reduces potentially harmful microorganisms on hands
  • Quickly and completely evaporates from the skin, leaving no residue
  • Packaged for dispensing through Fuzion Dispenser
  • Formulated without phosphates, butyl, or APE surfactants
  • Suitable for healthcare facilities, emergency services, offices, nursing homes, schools, and daycare centers as a supplement to regular hand washing.
Catalog No. 50-283-7599 Supplier MSC Supplier No. 52974557
Zep Instant Hand Sanitizer Gel is an alcohol-based hand sanitizer designed for effective hygiene without the need for water or towels. This 1200 mL gel helps to control many microorganisms of public health concern and can be used repeatedly throughout the day without irritation. It quickly and completely evaporates from the skin, leaving no residue, eliminating the need for towels.
  • May be used repeatedly without irritation
  • Reduces potentially harmful microorganisms on hands
  • Quickly and completely evaporates from the skin, leaving no residue
  • Packaged for dispensing through Fuzion Dispenser
  • Formulated without phosphates, butyl, or APE surfactants
  • Suitable for healthcare facilities, emergency services, offices, nursing homes, schools, and daycare centers as a supplement to regular hand washing.